Learn More
Apexbio Technology LLC AMI-1 20324-87-2 5mg
Supplier: Apexbio Technology LLC A19085
AMI-1 (CAS 20324-87-2) is a small molecule inhibitor of protein arginine N-methyltransferase 1 (PRMT1) exhibiting an IC50 of 8 8 M It acts by preventing the binding of peptide substrates to PRMT1 without interfering with the S-adenosyl-L-methionine (SAM) cofactor binding site In addition to its action on PRMT1 AMI-1 inhibits HIV-1 reverse transcriptase polymerase activity with an IC50 of 5 0 M AMI-1 is cell-permeable facilitating its use in cellular studies investigating arginine methylation epigenetic regulation and viral replication
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.