Learn More
Abcam AMI 1 Protein Arginine Methyltransferase Inhibitor, 99% Purity, Solid, 5 mg
Description
- Potency: Potent, selective and reversible protein arginine methyltransferase inhibitor (IC50 = 1.63 μM)
- Antioxidant Effect: ROS° scavenger with antioxidant effects (IC50 = 8.3 μM)
- Mechanism: Inhibits arginine in vitro without competing for the AdoMet binding site; modulates nuclear receptor-regulated transcription in vivo
- Form: Solid
- Purity: ≥99%
- Molecular Weight: 548.5 Da
- Permeability: Cell-permeable
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Protein Arginine Methyltransferase |
| Format | Vial |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Product Type | Antioxidant |
| Buffer | 100% disodium 7,7'-(carbonyldiimino)bis(4-hydroxynaphthalene-2-sulphonate) |
| Formulation | 100% Disodium 7,7'-(carbonyldiimino)bis(4-hydroxynaphthalene-2-sulphonate) |
| Quantity | 5 mg |
| For Use With (Application) | Protein arginine methyltransferase inhibition, ROS scavenging |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.