Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam AMI 1, Protein Arginine Methyltransferase Inhibitor, Solid, 25 mg

Catalog No. 502706586
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6586 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6586 Supplier Abcam Supplier No. AB14147225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective, and reversible protein arginine methyltransferase inhibitor with antioxidant effects.

  • Potent, selective and reversible protein arginine methyltransferase inhibitor (IC50 = 1.63 μM).
  • ROS° scavenger with antioxidant effects (IC50 = 8.3 μM).
  • Inhibits arginine in vitro without competing for the AdoMet binding site.
  • Modulates nuclear receptor-regulated transcription in vivo.
  • Cell-permeable.
  • Purity ≥99%.
  • MW: 548.5 Da.
  • Form: Solid.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule Protein Arginine Methyltransferase
Shipping Condition Ambient
Format Vial
Form Solid
Content And Storage Store at -20°C.
Product Type Antioxidant
Buffer 100% Disodium 7,7'-(carbonyldiimino)bis(4-hydroxynaphthalene-2-sulphonate)
Formulation 100% Disodium 7,7'-(carbonyldiimino)bis(4-hydroxynaphthalene-2-sulphonate)
Quantity 25 mg
For Use With (Application) Protein arginine methyltransferase inhibition, nuclear receptor-regulated transcription modulation, antioxidant research
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.