Learn More
MilliporeSigma™ Amicon™ Pro Affinity concentration Kit GST with Amicon™ Ultra-0.5 Device
Shop All MilliporeSigma ProductsDescription
Order Info
Reagents available. (See Cat. No. ACR5000NT)
Specifications
Specifications
| Color | Clear with Pink Cap |
| Material (Membrane) | Ultracel Low-binding Regenerated Cellulose |
| MWCO | 30 kDa |
| Filtration Area | 1 cm2 |
| For Use With (Application) | Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment |
| Housing Material | Polypropylene |
| Includes | 12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 30 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) |
| Max. Force | 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin |
| Quantity | 12/Pk. |
| Type | Amicon Pro Affinity Concentration Kit GST with 30 kd Amicon Ultra-0.5 Device |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.