Learn More
MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 30 Kda
Shop All MilliporeSigma ProductsDescription
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.
Specifications
Specifications
| Disposable | Single-use |
| MWCO | 30 kDa |
| Sterility | Non-sterile |
| Housing Material | Regenerated Cellulose (RC) |
| Includes | 24 Amicon Ultra NMWCO 30 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap |
| Quantity | 24Pack |
| Type | Centrifugal Filters |
| Volume (Metric) | 10 mL |
| Length (Metric) | 12 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.