Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Amicon™ Pro Purification System with Ultra-0.5 Device, 50 Kda

Catalog No. ACS505024 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
24Pack
Includes:
24 Amicon Ultra NMWCO 50 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Includes
ACS505024 24Pack 24 Amicon Ultra NMWCO 50 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACS505024 Supplier MilliporeSigma™ Supplier No. ACS505024

Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.

Amicon Pro Purification System - A Centrifugal Tool collapsing protein purification with concentration & buffer exchange.

The Amicon™ Pro Purification System includes: Amicon Ultra NMWCO filters, Exchange devices, and Collection tubes with cap.
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
MWCO 50 kDa
Sterility Non-sterile
Housing Material Regenerated Cellulose (RC)
Includes 24 Amicon Ultra NMWCO 50 KDa filters, 24 Exchange devices, and 24 Collection tubes with cap
Quantity 24Pack
Type Centrifugal Filters
Volume (Metric) 10 mL
Length (Metric) 12 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.