Learn More
Description
Includes
Two microcentrifuge tubes
- High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
- Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
- Heat-sealed membrane eliminates possibility of downstream extractables
- Universal rotor compatibility
- High retentate recovery of >90%
- High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
- Easy sample monitoring due to translucent housing and volume gradations
- Direct pipetter sample access eliminates processing step to recover concentrate
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates
- Concentration of dilute or prepurified proteins from column eluents
- Desalting and buffer exchange
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Disposable | Single-use |
| Material (Membrane) | High-recovery Ultracel-PL Regenerated Cellulose |
| MWCO | 30 kDa |
| Sterility | Non-sterile |
| Filtration Area | 1 cm2 |
| For Use With (Application) | Protein purification, protein concentration and desalting |
| Housing Material | Regenerated Cellulose (RC) |
| Includes | Two microcentrifuge tubes |
| Max. Force | 14,000 xG for concentration spin; 1000 xG for reverse spin |
| Quantity | 8Pack |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
