Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Amicon™ Ultra-4 Centrifugal Filter Units
GSA_VA

Catalog No. UFC800308 Shop All MilliporeSigma Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
Quantity:
24Pack
8 Pack
8Pack
96 Pack
96Pack
Diameter (Metric):
17.2 mm
17.3 mm
22.7 mm
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity Diameter (Metric)
UFC800308 3 kDa 8Pack 22.7 mm
UFC805008 50 kDa 8Pack 22.7 mm
UFC800324 3 kDa 24Pack 17.3 mm
UFC800396 3 kDa 96Pack 17.3 mm
UFC801008 10 kDa 8 Pack 22.7 mm
UFC801024 10 kDa 24Pack 17.2 mm
UFC801096 10 kDa 96 Pack 17.3 mm
UFC803008 30 kDa 8Pack 22.7 mm
UFC803024 30 kDa 24Pack 22.7 mm
UFC805024 50 kDa 24Pack 22.7 mm
UFC810008 100 kDa 8Pack 22.7 mm
UFC810024 100 kDa 24Pack 22.7 mm
UFC810096 100 kDa 96Pack 22.7 mm
UFC803096 30 kDa 96Pack 22.7 mm
UFC805096 50 kDa 96Pack 22.7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. UFC800308 Supplier MilliporeSigma™ Supplier No. UFC800308
Add to Cart
Add to Cart

Includes

Two microcentrifuge tubes

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material (Membrane) High-recovery Ultracel-PL Regenerated Cellulose
MWCO 3 kDa
Sterility Non-sterile
Filtration Area 3 cm2
For Use With (Application) Protein purification, protein concentration and desalting
Housing Material Regenerated Cellulose (RC)
Includes Two microcentrifuge tubes
Quantity 8Pack
Type Centrifugal Filters
Volume (Metric) 4 mL
Length (Metric) 124 mm
Diameter (Metric) 22.7 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.