Learn More
Enzo Life Sciences Amidepsine A (1mg). CAS: 169181-28-6
SDP
Supplier:  Enzo Life Sciences ALX380262M001
Encompass_Preferred
Inhibitor of diacylglycerol acyltransferase (DGAT). Shows inhibitory effect in rat liver microsomes (IC50=10.2µM) and Raji cells (IC50=15.5µM). Excessive accumulation of triacetylglycerol produced by DGAT may cause fatty liver, obesity and hypertriglyceridemia, which may lead to atherosclerosis, diabetes and metabolic disorders. Purity: ≥95% (HPLC). Appearance: Pale white to pale yellow powder. Solubility: Soluble in methanol, ethyl acetate or benzene; insoluble in water or hexane. Long Term Storage: -20°C.
Catalog No. 50-200-8993
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.