Learn More
Abcam Amido Black 10B Dye, Histological and Protein, Powder, 25 g
Description
- Molecular Weight: 616.5 g/mol
- Form: Powder
- Composition: 100% Naphthol Blue Black
- Applications: Diazo dye for protein staining on polyacrylamide gels, agarose gels, and nitrocellulose membranes; utilized in cell proliferation assays, histochemical studies, and as a forensic stain for enhancing latent prints contaminated with blood
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Protein |
| Shipping Condition | Ambient |
| Buffer | 100% Naphthol Blue Black |
| Form | Powder |
| Detection Method | Chromogenic |
| Membrane Compatibility | Nitrocellulose |
| Content And Storage | Store at ambient temperature. |
| Detection Location | In-Gel Detection, In-Blot Detection |
| Formulation | 100% Naphthol Blue Black |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.