Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Amido Black 10B, Histological and Protein Dye, Powder, 5 g

Catalog No. 502704735
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4735 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4735 Supplier Abcam Supplier No. AB1463115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Diazo dye for protein staining in gel electrophoresis, cell proliferation assays, and forensic blood-stain enhancement.

  • Molecular weight: 616.5 g/mol
  • Form: Powder
  • Composition: 100% Naphthol Blue Black
  • Applications: Staining proteins on polyacrylamide gels, agarose gels and nitrocellulose membranes; cell proliferation assays; histochemical studies; forensic stain for enhancing latent blood prints

Specifications

Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule Protein
Form Powder
Membrane Compatibility Nitrocellulose
Content And Storage Store at ambient temperature.
Detection Location In-Gel Detection, In-Blot Detection
Buffer 100% Naphthol Blue Black
Detection Method Colorimetric
Formulation 100% Naphthol Blue Black
Sample Type Polyacrylamide gels, Agarose gels, Nitrocellulose membranes, Blood-contaminated latent prints
Quantity 5 g
For Use With (Application) Protein staining, cell proliferation assays, histochemical studies, forensic stain for latent blood prints
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.