Learn More
Research Products International Corp Amikacin Disulfate 50 Grams
SDP
Supplier:  Research Products International Corp A2054050.0
Encompass_Preferred
Semisynthetic, aminoglycoside antibiotic derived from Kanamycin A. Range of activity is similar to Gentamicin. Active against many strains of Gram negative bacteria.
Catalog No. 50-238-8416
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.