Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Amine Reactive Comp-Bead 2 Population Kit
SDP

Catalog No. NB092254 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mL (2 x 5 mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092254 10 mL (2 x 5 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB092254 Supplier R&D Systems™ Supplier No. NBP300496

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. The Amine Reactive Compensation Beads are designed for labeling amine-reactive dyes (live/dead fixable viability stains) when setting compensation in flow cytometry.

Specifications

Formulation 0.016 M PBS (pH 7.4)
For Use With (Application) Flow Cytometry
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Line Novus
Quantity 10 mL (2 x 5 mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.