Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Amino Acid Diet, Synthetic, Pellet
Diet rich in Amino Acids. Can be added into agar gel to increase palatability.
Supplier: MP Biomedicals 0296036220
Description
- Diet rich in amino acids
- Can be added into agar gel to increase palatability
- Can be used for research

Specifications
Autoclavable | |
Single-use | |
20 kg | |
Refrigerated | |
0.3048 m |
Animal Diet | |
Pellet | |
Non-sterile | |
1 ft. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction