Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoacetaldehyde diethyl acetal, 99%, Thermo Scientific™

Catalog No. AC103070250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC103070250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC103070250 Supplier Thermo Scientific Chemicals Supplier No. 103070250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-36-3
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008136
InChI Key HJKLEAOXCZIMPI-UHFFFAOYSA-N
Synonym 2,2-diethoxyethylamine, aminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy, 2,2-diethoxyethan-1-amine, 2-aminoacetaldehyde diethyl acetal, glycinaldehyde diethyl acetal, aminoacetal, diethoxyethylamine, acetaldehyde, amino-, diethyl acetal, aminoacetaldehydediethylacetal
PubChem CID 69524
IUPAC Name 2,2-diethoxyethan-1-amine
SMILES CCOC(CN)OCC

Specifications

Density 0.9160g/mL
Boiling Point 162.0°C to 163.0°C
Flash Point 45°C
Refractive Index 1.4160 to 1.4180
Linear Formula H2NCH2CH(OC2H5)2
Quantity 25 mL
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, ether and chloroform
Formula Weight 133.19
Chemical Name or Material Aminoacetaldehyde diethyl acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.