Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aminoacetaldehyde dimethyl acetal

Catalog No. 121967500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-196-7500ML 500 mL
12-196-7100ML 100 mL
12-196-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-196-7500ML Supplier Sigma Aldrich Supplier No. 121967500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Aminoacetaldehyde dimethyl acetal reacts with sulfone, followed by hydrolysis and reductive amination by adding desired piperazine derivative to yield piperazine derivatives of 2-furanyl[1,2,4]triazolo[1,5-a][1,3,5]triazine.aminoacetaldehyde dimethyl acetal was used in a study to develop a fluorescent substrate for aldehyde dehydrogenase
  • It was used in preparation of chitosan-dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol)
  • It was used in an efficient 3-step synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by mgclo4 leading to α-aminophosphonates

Specifications

CAS 22483-09-6
Density 0.965 g/mL (at 25°C (literature))
Boiling Point 135°C to 139°C (95 mmHg)
Molecular Formula C4H11NO2
Refractive Index n20/D 1.417 (literature)
Linear Formula NH2CH2CH(OCH3)2
MDL Number MFCD00008135
Quantity 500 mL
Synonym 2,2-Dimethoxyethylamine
Molecular Weight (g/mol) 105.14
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.