Alfa Aesar™ Aminoacetonitrile sulfate, 97%

Manufacturer: Alfa Aesar™ B2215514

Quantity 25g

 View more versions of this product

Catalog No. AAB2215514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5466-22-8
    Molecular Formula C4H8N8·H2SO4
    Linear Formula (H2NCH2CN)2·H2SO4
    Formula Weight 210.21
    MDL Number MFCD00012851
    UN Number UN3439
    Beilstein 3916094
    Merck Index 14,412
    Synonym Glycinonitrile sulfate
    Chemical Name or Material Aminoacetonitrile sulfate

    Specifications

    Formula Weight 210.21
    Odor Odorless
    Merck Index 14,412
    Molecular Formula C4H8N8·H2SO4
    Linear Formula (H2NCH2CN)2·H2SO4
    MDL Number MFCD00012851
    CAS 5466-22-8
    UN Number UN3439
    Beilstein 3916094
    Melting Point ≈160°C (dec.)
    Synonym Glycinonitrile sulfate
    Quantity 25g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    RTECS Number MC2100000
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material Aminoacetonitrile sulfate
    Assay 97%

    Description

    Applications
    Aminoacetonitrile sulfate is used in organic synthesis, catalytic agent, petrochemical additive.

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Store away from strong oxidizing agents, water/ moisture.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit