Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoacetonitrile Sulfate 98.0+%, TCI America™
SDP

Catalog No. A091225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A091225G 25 g
A0912500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A091225G Supplier TCI America Supplier No. A091225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5466-22-8
Molecular Formula C2H4N2
MDL Number MFCD00012851
Synonym Cyanomethylamine Sulfate

Specifications

Melting Point 165°C
Color White
Quantity 25 g
UN Number 3439
Formula Weight 210.21
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Aminoacetonitrile Sulfate
EINECSNumber (2)-1519
RTECSNumber MC2100000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.