Learn More
Abcam Aminoguanidine hydrochloride, NOS inhibitor, 100MG
Supplier: Abcam AB120123100MG
MW 110.55 Da, Purity >98%. Irreversible inhibitor of iNOS, displaying 26-fold selectivity over nNOS. In vitro and in vivo inhibitor of the formation of glycosylation end products associated with diabetes and aging.
The product is subject to the following: Abcam Restricted Use Statement
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.