Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminoguanidine Sulfate Hydrate 98.0+%, TCI America™
SDP

Catalog No. A0309500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0309500G 500 g
A030925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0309500G Supplier TCI America Supplier No. A0309500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 996-19-0
Molecular Formula CH6N4
MDL Number MFCD00035665

Specifications

Melting Point 207°C
Color White
Quantity 500 g
Formula Weight 246.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Aminoguanidine Sulfate Hydrate
EINECSNumber (2)-1776&(9)-0070
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.