Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminomethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (0.5-0.9mmol/g), TCI America™
SDP

Catalog No. A154325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A154325G 25 g
A15435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A154325G Supplier TCI America Supplier No. A154325G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

MDL Number MFCD00130502

Specifications

Color Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Aminomethyl Polystyrene Resin cross-linked with 1% DVB (200-400mesh) (0.5-0.9mmol/g)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.