Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aminonaphtholsulfonic Acid Solution, Ricca Chemical
SDP

Catalog No. R06092004B Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R06092004B 4 L Black Poly Bottle
609-24 120 mL Amber Poly Bottle
609-216 500 mL Amber Poly Bottle
R06092001B 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R06092004B Supplier Ricca Chemical Company Supplier No. 609.21
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reducing Agent for Silica

Specifications

Color Colorless
CAS Min % 85.0, 12.71, 0.42, 0.21
CAS Max % 88.0, 13.23, 0.44, 0.22
Grade Laboratory
Packaging Black Poly Bottle
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Aminonaphtholsulfonic Acid Solution, Reducing Agent for Silica
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.