Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aminooxy-PEG3-bromide hydrobromide | 98.0% | 353.05 g·mol-1 | C8H19Br2NO4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY140396C5MG

Encompass_Preferred

Aminooxy-PEG3-bromide hydrobromide is a PEG-based bifunctional linker designed for use in the synthesis of proteolysis-targeting chimeras (PROTACs) and other bioconjugates. It combines an aminooxy reactive handle with a bromide-terminated PEG3 spacer to enable orthogonal coupling strategies in small-scale research workflows.

  • PEG-based linker suitable for PROTAC assembly.
  • Aminooxy group enables oxime ligation to aldehydes and ketones.
  • Bromide terminus allows alkylation or nucleophilic substitution reactions.
  • High purity (98.0%) supports reproducible synthetic results.
  • Available in small mg pack sizes for medicinal chemistry workflows.
  • Appearance: oil, convenient for solution-phase synthesis.

Catalog No. 50-002-23183


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.