Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amiodarone-D4 hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant

Catalog No. A0831ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0831ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0831ML Supplier MilliporeSigma Supplier No. A0831ML
Add to Cart
Add to Cart

A Certified Spiking Solution and stable-labeled internal standard for use in therapeutic drug monitoring analyses by LC/MS.

A Certified Spiking Solution™and stable-labeled internal standard for use in therapeutic drug monitoring analyses by LC/MS. Amiodarone, an antiarrhythmic agent marketed as Pacerone™, Cordarone™, Aratac, Atlansil, is monitored by clinical labs to ensure patients remain within the drug's therapeutic range.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amiodarone-D4 Hydrochloride
Packaging Ampule of 1 mL
CAS 1216715-80-8
Molecular Formula C25D4H26ClI2NO3
Concentration 100 μg/mL in Methanol (as free base)
For Use With (Equipment) Gas Chromatography (GC), Liquid Chromatography (LC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Clinical Testing
Molecular Weight (g/mol) 685.8
Quantity 1 mL
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.