Learn More
Cayman Chemical AmIselImodhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2097010 MG
Encompass_Preferred
A prodrug form of amiselimod phosphate; inhibits the development of chronic colitis induced by adoptive transfer of CD4+CD45RBhigh T cells in mice at 0.3 mg/kg per day
Catalog No. 50-298-2493