Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amitriptyline Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18600529 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-529 500 g Amber Glass Bottle
18-600-527 25 g Amber Glass Bottle
18-600-528 100 g Amber Glass Bottle
18-600-531 1 kg Amber Glass Bottle
18-600-530 10 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-529 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1322500GM
Add to Cart
Add to Cart

A1322, 549-18-8, C20H23N·HCl

Spectrum™ Chemical Amitriptyline Hydrochloride, USP is a medication of the tricyclic antidepressant class that is used for the treatment of mental illnesses. It is on the World Health Organization′s list of essential medicines. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 549-18-8
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.87
InChI Key KFYRPLNVJVHZGT-UHFFFAOYSA-N
IUPAC Name hydrogen dimethyl(3-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}propyl)amine chloride
SMILES [H+].[Cl-].CN(C)CCC=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 500 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.