Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amitriptyline Monoclonal Antibody (AM10-106.9)
GREENER_CHOICE

Catalog No. 01671840
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-840 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-840 Supplier Invitrogen™ Supplier No. MA110812
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10812 detects Amitriptyline. MA1-10812 has been successfully used in ELISA applications. The MA1-10812 immunogen is Amitriptyline, conjugated to KLH.

Amitriptyline is a Tricyclic Antidepressant that inhibits the re-uptake of norepinephrine and serotonin by the presynaptic neuronal membrane in the central nervous system (CNS), thereby increasing the synaptic concentration of norepinephrine and serotonin. Due to constant stimulation to these receptors, amitriptyline may produce a downregulation of adrenergic and serotonin receptors, which may contribute to the antidepressant activity. [National Center for Biotechnology Information. PubChem Compound Database; CID=2160].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amitriptyline
Applications ELISA
Classification Monoclonal
Clone AM10-106.9
Concentration ~1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C20H23N; Elatrol; Elavil; Endep; Laroxyl; Trepiline; Tryptanol; Tryptizol
Host Species Mouse
Immunogen Amitriptyline, conjugated to KLH.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.