Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amlitelimab Humanized Recombinant Human Monoclonal Antibody

Catalog No. MA554831
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554831 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA554831 Supplier Invitrogen™ Supplier No. MA554831
  • $423.00
    $317.65 / Each of 1

    Save $105.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

Amlitelimab is a monoclonal antibody that targets and inhibits OX40L, a protein involved in the activation and survival of T cells, thereby modulating the immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amlitelimab Humanized
Applications ELISA
Classification Recombinant Monoclonal
Concentration 2.9 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias TNFSF4; TXGP1
Host Species Human
Immunogen Recombinant Human CD252/TNFSF4.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG4 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.