Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amlodipine Besylate, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18600540 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-540 1 kg Amber Glass Bottle
18-600-537 5 g Amber Glass Bottle
18-600-538 25 g Amber Glass Bottle
18-600-539 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-540 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A39501KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A3950, 111470-99-6, C20H25ClN2O5·C6H6O3S

Spectrum™ Chemical Amlodipine Besylate, USP is a medication used to treat high blood pressure and coronary artery disease. It is on the World Health Organization's List of Essential Medicines. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 111470-99-6
Molecular Formula C26H31ClN2O8S
Molecular Weight (g/mol) 567.05
InChI Key ZPBWCRDSRKPIDG-UHFFFAOYNA-N
IUPAC Name 3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate; benzenesulfonic acid
SMILES OS(=O)(=O)C1=CC=CC=C1.CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Packaging Amber Glass Bottle
Quantity 1 kg
Optical Rotation -0.10° to +0.10°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.