accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOONH2NHClOOH3CCH3OHSOO

CAS RN 111470-99-6

IUPAC Name: 3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate; benzenesulfonic acid
Synonyms: amlodipine benzenesulfonate
Molecular Weight (g/mol): 567.05
Molecular Formula: C26H31ClN2O8S
InChi Key: ZPBWCRDSRKPIDG-UHFFFAOYNA-N
SMILES: OS(=O)(=O)C1=CC=CC=C1.CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC
Percent Purity 
  • (3)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)