Learn More
Abcam Ammonia Assay Kit, Fluorescent, Quantitative, 0.012 to 1 mM, 200 Test
Description
- Detection method: Fluorescent
- Range: 0.012 mM to 1 mM
- Results type: Quantitative
- Sample types: Urine, Other biological fluids
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Ammonia |
| No. of Reactions | 200 Tests |
| For Use With (Equipment) | Microplate |
| Format | Microplate Assay Kit |
| Detection Method | Fluorescence |
| Detection Location | In-Solution Detection |
| Sample Type | Urine, Other biological fluids |
| Quantity | 200 Tests |
| For Use With (Application) | Ammonia measurement in biofluids |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.