Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia ISE Fill Solution, Ricca Chemical
SDP

Catalog No. 3161200120A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3161200120A 120 mL
01-213-120 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3161200120A Supplier Ricca Chemical Company Supplier No. R3161200120A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 107-21-1 , 12125-02-9 , 7783-90-6
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.0, 0.54, 0.07, 0.01
CAS Max % 100.0, 0.56, 0.07, 0.01
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Ammonia ISE Fill Solution
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.