Learn More
Dentec Safety™ Ammonia and Methylamine Cartridge
Dentec Safety™ Ammonia and Methylamine Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity
Supplier: Dentec Safety™ 158T22
Description
- Offset connector allows the user to rotate the cartridge into 3 different positions improves visibility and allows face shields or welding helmets to be worn without interference
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Plastic | |
Masks | |
6/Box |
Respirator Accessories | |
Cartridges and filters |