Learn More
ILC Dover Ammonia & Multi-Hazard Cartridge Filters, Set of 3
Easy to use twist and lock Cartridge filters that protect against particulates, organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, and hydrogen fluoride.
Supplier:  ILC Dover S2027
Catalog No. 19-631-068
Only null left
Add to Cart
Description
Description
  • Easy to use twist and lock Cartridge filters
  • Protect against particulates, organic vapors, chlorine, hydrogen chloride, sulfur dioxide, chlorine dioxide, and hydrogen fluoride
Specifications
Specifications
Cartridge/Filter Combination
3/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.