Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia solution, 25.0%, Suprapur™, MilliporeSigma™

Catalog No. M1054280500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1054280500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1054280500 Supplier MilliporeSigma Supplier No. 1.05428.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym Ammonium hydroxide solution, Ammonia water
IUPAC Name amine hydrate
SMILES N.O

Specifications

Boiling Point 37.7°C (1013 hPa)
Melting Point −57.5°C
Assay Percent Range ≥ 25.0%
Density 0.903 g/cm3 (at 20°C)
Quantity 500 mL
Explosion Limit 15.4 to 33.6% (V)
Packaging Plastic Bottle
Vapor Pressure 483 hPa (20°C)
Recommended Storage Store at 2 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.