Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, 20% (w/v), Ricca Chemical
SDP

Catalog No. R06198001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0619800-1A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R0619800-1A Supplier Ricca Chemical Company Supplier No. R06198001A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Color Colorless
CAS Min % 79.0, 19.0
CAS Max % 81.0, 21.0
Physical Form Liquid
Quantity 1 L
Percent Purity 20% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 20% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Ammonium Acetate
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.