Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, 65% w/v, Reagents

Catalog No. 01336122
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-122 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-122 Supplier Reagents Holdings Llc Supplier No. CA0063004A
Only null left
Add to Cart
Add to Cart

Ammonium Acetate, 65% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 631-61-8 , 7732-18-5
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.08
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
IUPAC Name ammonium acetate
SMILES [NH4+].CC([O-])=O

Specifications

Boiling Point 109°C
Color Colorless
Physical Form Liquid
Quantity 4 L
Percent Purity 65%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.08
Chemical Name or Material Ammonium Acetate, 65% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.