Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium acetate, ACS, 97.0% min

Catalog No. p-6827151
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1159936 500 g
AA11599A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1159936 Supplier Thermo Scientific Chemicals Supplier No. 01159936
Only null left
Add to Cart

Reagent in analytical chemistry, drugs | CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol

Ammonium acetate is a reagent used in chromatographic analysis of various compounds such as oligos, proteins, and peptides. It is also employed with acetic acid to provide a buffer solution. Generally it behaves as a catalyst in many reactions like Knoevenagel, Borch. It is also employed as a nutrient additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point ∽114°C (decomposition)
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 97.0% min
Beilstein 4186741
Flash Point 136°C (277°F)
Solubility Information Soluble in cold water,methanol ,SO2and acetone.
Formula Weight 77.08
Grade ACS
Sensitivity Hygroscopic
Density 1.17
Chemical Name or Material Ammonium acetate
Show More Show Less
EINECSNumber 211-162-9
RTECSNumber AF3675000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.