Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, ACS Grade, ≥ 97%, LabChem™

Catalog No. LC109601 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC109601 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC109601 Supplier LabChem, Inc. Supplier No. LC109601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula CH3COONH4
Flash Point 136°C
Solubility Information Soluble in water
Formula Weight 77.08
Percent Purity ≥97%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Gaseous Ammonia; Carbon dioxide; Carbon monoxide; Nitrogen oxides
Chemical Name or Material Ammonium Acetate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Avoid breathing dust, vapors.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Keep container tightly closed.
Store locked up and in a well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.