Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate Buffer, For Iron, APHA 3500-Fe B, Certified, LabChem™

Catalog No. LC109702
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC109702 1 L
LC109701 500 mL
LC109704 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC109702 Supplier LabChem, Inc. Supplier No. LC109702
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Ammonium Acetate Buffer
Name Note For Iron
CAS 64-19-7, 631-61-8, 7732-18-5
Color Colorless
CAS Max % 68.75, 22.36, 8.89
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors; Gaseous Ammonia
Physical Form Liquid
Quantity 1 L
Identification Passes Test
Molecular Formula CH3COONH4
Linear Formula CH3COONH4
Packaging Poly Bottle
UN Number UN2790
Solubility Information Soluble in water
Formula Weight 77.08
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.