Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, Crystal, ACS, 97%, Spectrum™ Chemical
SDP

Catalog No. 18608574 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-574 500 g Poly Bottle
18-608-572 125 g Poly Bottle
18-608-569 2.5 kg Poly Bottle
18-608-573 12 kg Poly Bottle
18-608-570 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-574 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1120500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.08
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
IUPAC Name ammonium acetate
SMILES [NH4+].CC([O-])=O

Specifications

pH 6.7 to 7.3
CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 500 g
Percent Purity 97%
Grade Reagent
Identification To Pass Test
Packaging Poly Bottle
Residue after Ignition 0.0001
Chemical Name or Material Ammonium Acetate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.