Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1011169050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Carton
Glass Bottle
Plastic Bottle
Plastic Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1011169050 50 kg Fiber Carton
M1011160500 500 g Glass Bottle
M1011161000 1 kg Plastic Bottle
M1011169012 12 kg Plastic Drum
M1011169025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. M1011169050 Supplier MilliporeSigma Supplier No. 1011169050
Add to Cart
Add to Cart
Request a Quote

Recommended Storage

+15°C to +25°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Melting Point 114°C
Color Colorless
pH 6.0 to 7.5
Physical Form Solid
Quantity 50 kg
Solubility Information 1480g/L
Percent Purity >96%
Packaging Fiber Carton
Density 1.17 g/3 (20 °C)
EINECSNumber 211-162-9
RTECSNumber AF3675000
Recommended Storage +15°C to +25°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.