Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Acetate, Purified, Spectrum™ Chemical
SDP

Catalog No. 18608820 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-820 25 g Poly Pail
18-608-818 500 g Poly Pail
18-608-819 2.5 kg Poly Pail
18-608-816 12 kg Poly Pail
18-608-817 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-820 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A112125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 631-61-8

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 25 g
Molecular Weight (g/mol) 77.08
Grade Purified
Packaging Poly Pail
Chemical Name or Material Ammonium Acetate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.