Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Aluminum Sulfate Dodecahydrate, Reagent Grade, ≥99% (Titration), Solstice

Catalog No. 60047816 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-816 1 kg Poly Bottle
60-047-817 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-816 Supplier Solstice Supplier No. A21401KG
Add to Cart
Add to Cart

Reagent Grade,≥99% (titration)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-26-1
Molecular Formula AlH28NO20S2
Molecular Weight (g/mol) 453.313
MDL Number MFCD00149958
InChI Key WZUKKIPWIPZMAS-UHFFFAOYSA-K
Synonym ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate
PubChem CID 62668
IUPAC Name aluminum;azanium;disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3]

Specifications

Melting Point 93.5°C
Color White
pH 2.6 (at 25°C)
Quantity 1 kg
Linear Formula AlNH4(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 453.33g/mol
Percent Purity ≥99% (titration)
Odor Odorless
Packaging Poly Bottle
Appearance Crystalline
Density 1.640g/cm3 at 20°C
Chemical Name or Material Ammonium aluminum sulfate dodecahydrate
Show More Show Less
RTECSNumber 232-055-3
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.