Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bicarbonate, (Crystalline),MP Biomedicals

Catalog No. ICN15010791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15010791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15010791 Supplier MP Biomedicals, Inc Supplier No. 0215010791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium bicarbonate for research use only.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

Physical Form Crystalline
Quantity 1 kg
Formula Weight 79.1
Percent Purity 99.65%
Chemical Name or Material Ammonium Bicarbonate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.