CAS RN 1066-33-7
Ammonium bicarbonate, 99%, for analysis
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium Bicarbonate (Powder/Certified), Fisher Chemical™
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium hydrogen carbonate, 99.0%
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium bicarbonate, 98%, pure
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium Bicarbonate, (Crystalline),MP Biomedicals
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium hydrogen carbonate, 98%
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
0.5% 1M Ammonium Bicarbonate in Methanol, Solstice
LabReady™ Solvent Blend, Custom Reagent
Ammonium Bicarbonate Crystalline MP Biomedicals
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Methanol 90% with 50mM Ammonium Bicarbonate, LabReady™ Solvent Blend, Solstice
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium Bicarbonate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium Bicarbonate, 0.1 M, Ricca Chemical
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium bicarbonate, Solstice
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Ammonium bicarbonate, Puriss., meets analytical spec. of Ph.Eur., BP, E 503, 99-101%, Solstice
CAS: 1066-33-7 Molecular Formula: CH5NO3 Molecular Weight (g/mol): 79.06 MDL Number: MFCD00012138 InChI Key: ATRRKUHOCOJYRX-UHFFFAOYSA-N Synonym: ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327 PubChem CID: 14013 IUPAC Name: carbonic acid amine SMILES: N.OC(O)=O
Fisher Scientific Ammonium Bicarbonate, Fisher BioReagents
CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol