Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium bicarbonate, Puriss., meets analytical spec. of Ph.Eur., BP, E 503, 99-101%, Solstice

Catalog No. 60044691 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-691 6 x 1 kg Poly Bottle
60-019-02 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-691 Supplier Solstice Supplier No. 112136X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Physical Form Powder
Quantity 6 x 1 kg
Assay Percent Range 99 to 101%
Linear Formula NH4HCO3
Beilstein 4329606
Formula Weight 79.06
Percent Purity 99 to 101%
Grade puriss.
Packaging Poly Bottle
Density 1.58 g/cm3; Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium bicarbonate, Puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%
Show More Show Less
Hazard Statement P301 + P312 + P330
Precautionary Statement H302
ShelfLife 1260 days from date of manufacture
EINECSNumber 213-911-5
RTECSNumber BO8600000
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.