Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Bifluoride, Purified, Reagents

Catalog No. 01337503
Change view
Click to view available options
Quantity:
100 mL
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-503 3 kg
01-337-506 100 mL
01-337-502 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-503 Supplier Reagents Holdings Llc Supplier No. C21305003A5
Add to Cart
Add to Cart

Ammonium Bifluoride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium Hydrogen Difluoride, Ammonium Fluoride Hydrofluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 240°C
Melting Point 124.6°C
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.5
Chemical Name or Material Ammonium Bifluoride, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.