Learn More
Ammonium carbamate, 98%
Description
Ammonium carbamate is used in the preparation of fertilizers. It acts as an ammoniating agent. It plays an important role in the formation of carbamoyl phosphate, which is essential for urea cycle and pyrimidines production. In addition to this, it is used in the preparation of trimethylsilyl carbamate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 60° (sublimation) |
| Physical Form | Powder |
| Quantity | 50 g |
| Beilstein | 3914091 |
| Solubility Information | Soluble in cold water and ammonium hydroxide. Slightly soluble in alcohol. Insoluble in acetone. |
| Formula Weight | 78.07 |
| Density | 1.6 |
| Chemical Name or Material | Ammonium carbamate |
Safety and Handling
| Hazard Category | H302-H318 |
| Precautionary Statement | P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c |
| EINECSNumber | 214-185-2 |
| RTECSNumber | EY8575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.