Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Carbonate
SCI_ED

Catalog No. S25167 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Chips
Lumps
Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25167 500 g Powder
S25166 100 g Chips
S25166A 500 g Lumps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25167
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Color Colorless to White
Quantity 500 g
Linear Formula (NH4)2CO3
Formula Weight 96.07
Grade Laboratory
Packaging Powder
Density 1.5g/mL at 20°C
Chemical Name or Material Ammonium Carbonate
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.